missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride hexahydrate, 98+%, extra pure

Catalog No. p-4813526
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC447700010 1 kg
AC447700050 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447700010 Supplier Thermo Scientific Chemicals Supplier No. 447700010
Only null left

CAS: 7791-18-6 | Cl2H12MgO6 | 203.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Melting Point 117.0°C
Quantity 1 kg
Assay Percent Range 98.0 to 101% (Complexometry)
Linear Formula MgCl2·6H2O
Merck Index 15,5726
Solubility Information Solubility in water: 2350g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 203.31
Percent Purity ≥98%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Magnesium chloride hexahydrate
Show More Show Less

RUO – Research Use Only