missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride, 6-Hydrate, Crystal, U.S.P. - F.C.C., J.T. Baker™

Catalog No. p-5390780
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003262 500 g Poly Bottle
02003261 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02003262 Supplier Avantor Supplier No. 244801
Only null left

Chemical Identifiers

CAS 7791-18-6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Physical Form Crystals
Quantity 500 g
Molecular Formula MgCl2·6H2O
Formula Weight 203.3
Grade FCC/USP
Packaging Poly Bottle
Specific Gravity 1L = 1.57kg
Chemical Name or Material Magnesium Chloride, 6-Hydrate