missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Magnesium chloride

Catalog No. 503124502
Encompass
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
503124502 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 503124502 Supplier Sigma Aldrich Supplier No. M82661KG

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Magnesium chloride

Specifications

CAS 7786-30-3
Melting Point 714°C
Density 2.32 g/mL (at 25°C)
Boiling Point 1412°C
Molecular Formula Cl2Mg
Linear Formula MgCl2
Quantity 1 kg
MDL Number MFCD00011106
Synonym Magnogene
Molecular Weight (g/mol) 95.21
Percent Purity ≥98%
Show More Show Less