missing translation for 'onlineSavingsMsg'
Learn More

Magnesium carbonate basic, heavy, extra pure

Magnesium carbonate basic, heavy, 40 to 45% (MgO), CMgO3, CAS Number-39409-82-0, 500g, loss on calcination: 55 to 60%, White, 208-915-9, 84.31, Extra Pure, MgCO3, 15,5724, MFCD00011107, Plastic bottle, Powder, OM2470000 | CAS: 39409-82-0 | C4H10Mg5O18 | 467.63 g/mol

$230.48 - $510.51

Chemical Identifiers

CAS 39409-82-0
Molecular Formula C4H10Mg5O18
Molecular Weight (g/mol) 467.63
MDL Number MFCD00149783
InChI Key YMKSTRLFWJQKFP-UHFFFAOYSA-D
IUPAC Name pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
SMILES O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Qty  
Catalog Number Mfr. No. Quantity Packaging Price Qty  
AC413405000
SDS View Documents
Thermo Scientific Chemicals
413405000
500 g Plastic bottle
Each for $230.48
Only null left
 
AC413400025
SDS View Documents
Thermo Scientific Chemicals
413400025
2.5 kg Plastic Drum
Each for $510.51
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

39409-82-0
467.63
YMKSTRLFWJQKFP-UHFFFAOYSA-D
O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
C4H10Mg5O18
MFCD00149783
pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate

Specifications

White
Powder
C4H10Mg5O18
MFCD00149783
Solubility in water: insoluble. Other solubilities: insoluble in ethanol, soluble in acids
O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
467.63
40 to 45% (MgO)
Plastic bottle
39409-82-0
500 g
MgCO3
15,5724
YMKSTRLFWJQKFP-UHFFFAOYSA-D
pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
84.31
Extra Pure
Magnesium carbonate basic, heavy
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-915-9

RTECSNumber : OM2470000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only