missing translation for 'onlineSavingsMsg'
Learn More

Magnesium carbonate basic, heavy, extra pure

Catalog No. AC413405000
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Plastic Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413405000 500 g Plastic bottle
AC413400025 2.5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC413405000 Supplier Thermo Scientific Chemicals Supplier No. 413405000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Magnesium carbonate basic, heavy, 40 to 45% (MgO), CMgO3, CAS Number-39409-82-0, 500g, loss on calcination: 55 to 60%, White, 208-915-9, 84.31, Extra Pure, MgCO3, 15,5724, MFCD00011107, Plastic bottle, Powder, OM2470000 | CAS: 39409-82-0 | C4H10Mg5O18 | 467.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39409-82-0
Molecular Formula C4H10Mg5O18
Molecular Weight (g/mol) 467.63
MDL Number MFCD00149783
InChI Key YMKSTRLFWJQKFP-UHFFFAOYSA-D
IUPAC Name pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
SMILES O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 500 g
Packaging Plastic bottle
Linear Formula MgCO3
Merck Index 15,5724
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in ethanol, soluble in acids
Formula Weight 84.31
Percent Purity 40 to 45% (MgO)
Grade Extra Pure
Chemical Name or Material Magnesium carbonate basic, heavy
Show More Show Less
EINECSNumber 208-915-9
RTECSNumber OM2470000
TSCA TSCA

RUO – Research Use Only