missing translation for 'onlineSavingsMsg'
Learn More

Magnesium bromide, 98%, pure, anhydrous

Catalog No. AC413390050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413390050 5 g Glass bottle
AC413390010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC413390050 Supplier Thermo Scientific Chemicals Supplier No. 413390050
Only null left

CAS: 7789-48-2 | Br2Mg | 184.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-48-2
Molecular Formula Br2Mg
Molecular Weight (g/mol) 184.11
MDL Number MFCD27995637
InChI Key OTCKOJUMXQWKQG-UHFFFAOYSA-L
Synonym magnesium bromide, magnesium dibromide, dibromomagnesium, magnesium-bromide, acmc-20aklq, magnesium bromide salt, magnesium ii bromide, magnesium 2+ ion dibromide, grignard reagent
PubChem CID 82241
IUPAC Name magnesium;dibromide
SMILES [Mg++].[Br-].[Br-]

Specifications

Melting Point 711.0°C
Quantity 5 g
Assay Percent Range 97.5 to 102.5% (Argentometry)
Linear Formula MgBr2
Fieser 02,254; 06,270; 08,273; 10,14; 15,194; 16,199
Merck Index 15,5723
Solubility Information Solubility in water: 1015g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 184.11
Percent Purity 98%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Magnesium bromide, Anhydrous
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 232-170-9

RUO – Research Use Only