missing translation for 'onlineSavingsMsg'
Learn More

Magnesium bis(trifluoromethylsulfonyl)imide

Catalog No. p-7080173
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA46667MD 250 mg
AA4666703 1 g
AA4666706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA46667MD Supplier Thermo Scientific Chemicals Supplier No. 046667MD
Only null left

CAS: 133395-16-1 | C4F12MgN2O8S4 | 584.576 g/mol

Magnesium bis(trifluoromethylsulfonyl)imide is a strong Lewis acid and used as a catalyst in organic synthesis. It is an electrolyte, which shows high anodic stability used in rechargeable magnesium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 133395-16-1
Molecular Formula C4F12MgN2O8S4
Molecular Weight (g/mol) 584.576
MDL Number MFCD01321278
InChI Key DMFBPGIDUUNBRU-UHFFFAOYSA-N
Synonym bis bis trifluoromethylsulfonyl amino magnesium, magnesium bis trifluoromethanesulfonimide, magnesium ii bis trifluoromethanesulfonyl imide, magnesium ii triflimide, magnesium bistrifluoromethanesulfonimide, magnesium bis trifluoromethylsulfonyl imide, magnesium 2+ bis bistriflylimide anion, bis trifluoromethanesulfonyl imide magnesium ii salt
PubChem CID 11851070
IUPAC Name magnesium;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Mg+2]

Specifications

Melting Point >200°C
Color White to Yellow
Linear Formula [(CF3SO2)2N]2Mg
Quantity 250 mg
UN Number UN1759
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 584.58
Physical Form Solid
Chemical Name or Material Magnesium bis (trifluoromethylsulfonyl)imide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only