missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Aluminum Silicate, Type IA, NF, Spectrum™ Chemical

Catalog No. 18604011
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604011 500 g Poly Pail
18604012 2.5 kg Poly Pail
18604013 12 kg Fiber Drum
18604014 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604011 Supplier Spectrum Chemical Mfg Cor Supplier No. M1021500GM

M1021, 1327-43-1

Spectrum™ Chemical Magnesium Aluminum Silicate, Type IA, NF is used in the pharmaceutical formulation process as an absorbent, anticaking agent, opacifying agent, slip modifier, and an aqueous viscosity increasing agent. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1327-43-1

Specifications

pH 9.0-10.0
CAS Max % 100%
Quantity 500 g
Grade NF
Loss on Drying 8%
Packaging Poly Pail