missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Acetate Tetrahydrate, Solstice

Catalog No. 6002157
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002157 500 g
6002156 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002157 Supplier Solstice Supplier No. 228648500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 16674-78-5
Molecular Formula C4H14MgO8
Molecular Weight (g/mol) 214.45
MDL Number MFCD00149214
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
Synonym magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
PubChem CID 134717
IUPAC Name magnesium;diacetate;tetrahydrate
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O

Specifications

Physical Form Solid
Quantity 500 g
Linear Formula (CH3COO)2Mg · 4H2O
UN Number NONH for all modes of transport
Beilstein 3730605
Formula Weight 214.45g/mol
Percent Purity ≥98%
Grade ACS Reagent
Chemical Name or Material Magnesium Acetate Tetrahydrate
EINECSNumber 205-554-9
RTECSNumber AI5600000