missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Magnesium acetate tetrahydrate, for biochemistry

Chemical Identifiers

CAS 16674-78-5
Molecular Formula C4H14MgO8
Molecular Weight (g/mol) 214.45
MDL Number MFCD00149214
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
Synonym magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
PubChem CID 134717
IUPAC Name magnesium;diacetate;tetrahydrate
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC212552500
SDS View Documents
Thermo Scientific™
212552500
250g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC212550010
SDS View Documents
Thermo Scientific™
212550010
1kg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For biochemistry
Specifications

Chemical Identifiers

16674-78-5
214.45
XKPKPGCRSHFTKM-UHFFFAOYSA-L
134717
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
C4H14MgO8
MFCD00149214
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
magnesium;diacetate;tetrahydrate

Specifications

16674-78-5
C4H14MgO8
MFCD00149214
15,5718
Solubility in water: 1200g/L (15°C). Other solubilities: soluble in alcohol
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
214.45
214.46
Plastic bottle
250g
(CH3CO2)2Mg·4H2O
15,196
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
XKPKPGCRSHFTKM-UHFFFAOYSA-L
magnesium;diacetate;tetrahydrate
134717
Biochemistry
Magnesium acetate tetrahydrate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only