missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Magnesium acetate tetrahydrate, for biochemistry

Catalog No. AC212552500
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC212552500 250 g
AC212550010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC212552500 Supplier Thermo Scientific Chemicals Supplier No. 212552500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16674-78-5 | C4H14MgO8 | 214.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For biochemistry

Chemical Identifiers

CAS 16674-78-5
Molecular Formula C4H14MgO8
Molecular Weight (g/mol) 214.45
MDL Number MFCD00149214
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
Synonym magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
PubChem CID 134717
IUPAC Name magnesium;diacetate;tetrahydrate
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 80.0°C
Quantity 250 g
Packaging Plastic bottle
Linear Formula (CH3CO2)2Mg·4H2O
Fieser 15,196
Merck Index 15,5718
Solubility Information Solubility in water: 1200g/L (15°C). Other solubilities: soluble in alcohol
Formula Weight 214.46
Grade Biochemical
Chemical Name or Material Magnesium acetate tetrahydrate

RUO – Research Use Only