missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Acetate Tetrahydrate, ACS Reagent, Reagents

Catalog No. 01337917
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337917 1 kg
01337916 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337917 Supplier Labchem Inc Supplier No. C12376001KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Magnesium Acetate Tetrahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 16674-78-5
Molecular Formula C4H14MgO8
Molecular Weight (g/mol) 214.45
MDL Number MFCD00149214
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
Synonym Magnesium diacetate tetrahydrate
IUPAC Name magnesium(2+) diacetate tetrahydrate
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 1 kg
Grade ACS
Packaging Poly Bottle
Specific Gravity 1.45
Chemical Name or Material Magnesium Acetate Tetrahydrate, ACS Reagent