missing translation for 'onlineSavingsMsg'
Learn More

Magnesium acetate tetrahydrate, 99.5%, for analysis

CAS: 16674-78-5 | C4H14MgO8 | 214.45 g/mol

Supplier:  Thermo Scientific Chemicals 270500010

 View more versions of this product

Catalog No. AC270500010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
Specifications

Specifications

Magnesium acetate tetrahydrate
p.a.
White
Crystalline Powder
99.5% min. (Complexometry)
(CH3CO2)2Mg·4H2O
15,196
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
XKPKPGCRSHFTKM-UHFFFAOYSA-L
magnesium(2+) diacetate tetrahydrate
134717
99.5%
Authentic
80.0°C
16674-78-5
8 to 9 (5% soln. at 20°C)
1 kg
C4H14MgO8
MFCD00149214
15,5718
Solubility in water: 1200g/L (15°C). Other solubilities: soluble in alcohol
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
214.45
214.46
Analytical
Plastic Bottle

Chemical Identifiers

16674-78-5
214.45
XKPKPGCRSHFTKM-UHFFFAOYSA-L
134717
C4H14MgO8
MFCD00149214
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only