missing translation for 'onlineSavingsMsg'
Learn More

Magnesium acetate tetrahydrate, 98% (metals basis)

Catalog No. p-7048966
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
250 g
Catalog No. Quantity
AA1222530 250 g
AA12225A1 1 kg
2 options

Catalog No. AA1222530

Supplier: Thermo Scientific Chemicals 01222530

Only null left

CAS: 16674-78-5 | C4H14MgO8 | 214.45 g/mol

Magnesium acetate tetrahydrate is commonly used as a source of magnesium in biological reactions It is an essential compound for a number of biochemical processes. Involved in glycolysis and the Krebs cycle, formation of cyclic-AMP and ATP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16674-78-5
Molecular Formula C4H14MgO8
Molecular Weight (g/mol) 214.45
MDL Number MFCD00149214
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
Synonym magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
PubChem CID 134717
IUPAC Name magnesium;diacetate;tetrahydrate
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 80°C
Physical Form Crystalline
Quantity 250 g
Beilstein 3730605
Merck Index 14,5654
Solubility Information Freely soluble in water, alcohol
Formula Weight 214.45 (142.39 Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 1.454 g/mL
Chemical Name or Material Magnesium acetate tetrahydrate
Show More Show Less
Hazard Category H500
EINECSNumber 205-554-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only