missing translation for 'onlineSavingsMsg'
Learn More

Magnesium acetate tetrahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

$41.72 - $269.98

Chemical Identifiers

CAS 16674-78-5
Molecular Formula C4H14MgO8
Molecular Weight (g/mol) 214.45
MDL Number MFCD00149214
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
Synonym magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
PubChem CID 134717
IUPAC Name magnesium(2+) diacetate tetrahydrate
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC423870050
SDS View Documents
Thermo Scientific Chemicals
423870050
5 g Glass bottle
Each for $41.72
Only null left
 
AC423875000
SDS View Documents
Thermo Scientific Chemicals
423875000
500 g Plastic bottle
Each for $269.98
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16674-78-5
214.45
XKPKPGCRSHFTKM-UHFFFAOYSA-L
134717
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
C4H14MgO8
MFCD00149214
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
magnesium(2+) diacetate tetrahydrate

Specifications

80.0°C
White
5 g
C4H14MgO8
MFCD00149214
15, 5718
Solubility in water: 1200g/L (15°C). Other solubilities: soluble in alcohol
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
214.45
214.46
ACS Reagent
Glass bottle
16674-78-5
Crystalline Powder and/or Crystals
98.0 to 102% (Complexometry)
(CH3CO2)2Mg·4H2O
15,196
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
XKPKPGCRSHFTKM-UHFFFAOYSA-L
magnesium(2+) diacetate tetrahydrate
134717
≥98%
Authentic
Magnesium acetate tetrahydrate
Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only