missing translation for 'onlineSavingsMsg'
Learn More

Magnesium acetate tetrahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog No. AC423875000
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423875000 500 g Plastic bottle
AC423870050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC423875000 Supplier Thermo Scientific Chemicals Supplier No. 423875000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16674-78-5 | Molecular Formula: C4H14MgO8 | Molecular Weight: 214.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16674-78-5
Molecular Formula C4H14MgO8
Molecular Weight (g/mol) 214.45
MDL Number MFCD00149214
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
Synonym magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
PubChem CID 134717
IUPAC Name magnesium(2+) diacetate tetrahydrate
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 80.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Assay Percent Range 98.0 to 102% (Complexometry)
Linear Formula (CH3CO2)2Mg·4H2O
Fieser 15,196
Merck Index 15, 5718
Solubility Information Solubility in water: 1200g/L (15°C). Other solubilities: soluble in alcohol
Formula Weight 214.46
Percent Purity ≥98%
Grade ACS Reagent
Chemical Name or Material Magnesium acetate tetrahydrate
Show More Show Less
Hazard Category HYGROSCOPIC

RUO – Research Use Only