missing translation for 'onlineSavingsMsg'
Learn More

Magnesium acetate tetrahydrate, 97.5%, extra pure, crystalline

CAS: 16674-78-5 | C4H14MgO8 | 214.45 g/mol

$110.82 - $110.82

Chemical Identifiers

CAS 16674-78-5
Molecular Formula C4H14MgO8
Molecular Weight (g/mol) 214.45
MDL Number MFCD00149214
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
Synonym magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
PubChem CID 134717
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC211572500
SDS View Documents
Thermo Scientific Chemicals
211572500
250 g Plastic bottle
Each for $110.82
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16674-78-5
214.45
XKPKPGCRSHFTKM-UHFFFAOYSA-L
134717
C4H14MgO8
MFCD00149214
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O

Specifications

80.0°C
White
250 g
C4H14MgO8
MFCD00149214
15, 5718
Solubility in water: 1200g/L (15°C). Other solubilities: soluble in alcohol
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
214.45
214.46
Extra Pure
Plastic bottle
16674-78-5
Crystalline Powder
97% min. (Complexometry)
(CH3CO2)2Mg·4H2O
15,196
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
XKPKPGCRSHFTKM-UHFFFAOYSA-L
magnesium;diacetate;tetrahydrate
134717
97.5%
Authentic
Magnesium acetate tetrahydrate, Extra Pure
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only