missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MagnaBind™ Goat Anti-Mouse IgG

Catalog No. PI21354
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21354 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21354 Supplier Thermo Scientific™ Supplier No. LSG21354

missing translation for 'validMainframeMsg'

Enable benchtop magnetic affinity purification with these ligand-coated (streptavidin, Protein A and G, GAM IgG, GAR IgG) magnetic particles and magnet separators.

Thermo Scientific™ MagnaBind™ Beads provide a convenient method for magnetic separation of antibodies, antigens, lectins, enzymes, nucleic acids and cells using affinity binding. To remove the MagnaBind Beads from the suspension, an external magnetic field is used.

MagnaBind Goat Anti-Mouse IgG Beads can be used to conveniently separate cells of interest from a cell mixture. The monoclonal or polyclonal antibody to the cell surface antigen is pre-incubated with the appropriate magnetic bead that is then incubated with the cell suspension.

Features of MagnaBind Goat Anti-Mouse IgG Beads:

  • Composition: Silanized iron oxide
  • Magnetization: 25-35EMU/g
  • Type of Magnetization: Superparamagnetic (no magnetic memory)
  • Surface Area: >100m2/g
  • Bead Size: 1-4μm diameter
  • Settling Rate: 4% in 30 minutes
  • Effective Density: 2.5g/mL
  • Number of Beads: 1 x 108 beads/mg
  • pH Stability: Aqueous solution, above pH 4.0
  • Concentration: ∼1 mg/mL

Applications:

  • Cell sorting using either positive or negative selection
  • Protein purification or immunoassays using either direct or indirect methods

Specifications

Concentration Slurry: ∼1 mg/mL
Content And Storage Upon receipt store products at 4°C. Do not freeze products. Products are shipped at ambient temperature.
Description MagnaBind Goat Anti-Mouse IgG Beads
Color Brown
For Use With (Application) Affinity Purification
Product Type Antibody Coated Bead
Ligand Type Antibody
Product Line MagnaBind
Quantity 50 mL
Volume (Metric) 50 mL
Diameter (Metric) 1 to 4 μm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.