missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MagnaBind™ Streptavidin Beads

Catalog No. PI21344
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21344 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21344 Supplier Thermo Scientific™ Supplier No. 21344

Enable benchtop magnetic affinity purification with these ligand-coated (streptavidin, Protein A and G, GAM IgG, GAR IgG) magnetic particles and magnet separators.

Thermo Scientific MagnaBind Streptavidin Beads are convenient for affinity purification or separation methods involving of biotin-labeled molecules. To remove the MagnaBind Beads from the suspension, an external magnetic field is used.

Features of MagnaBind Streptavidin Beads:

  • Composition: Silanized iron oxide
  • Magnetization: 25-35EMU/g
  • Type of Magnetization: Superparamagnetic (no magnetic memory)
  • Surface Area: >100m2/g
  • Bead Size: 1-4μm diameter
  • Settling Rate: 4% in 30 minutes
  • Effective Density: 2.5g/mL
  • Number of Beads: 1 x 108 beads/mg
  • pH Stability: Aqueous solution, above pH 4.0
  • Concentration: 5 mg/mL

Streptavidin is a 60kDa protein from Streptomycetes avidinii. The protein is a tetramer having four biotin-binding sites. Unlike avidin, streptvidin has a low isoelectric point (pI=5) and no carbohydrate groups, resulting in low nonspecific binding. The high-affinity interaction between streptavidin and biotin cannot be dissociated efficiently except with very harsh conditions, such as boiling in sample loading buffer for SDS-PAGE or 8M guanidine HCl, pH 1.5. Consequently, it is often possible to elute binding partners in an interaction complex without also eluting the biotinylated component.

Applications:

  • Cell sorting using either positive or negative selection
  • Protein purification or immunoassays using either direct or indirect methods

Specifications

Concentration Slurry: 5 mg/mL
Content And Storage Upon receipt store product at 4°C. Do not freeze product. Product is shipped at ambient temperature.
Description MagnaBind Streptavidin Beads
Color Brown
For Use With (Application) Affinity Purification
Product Type Streptavidin Magnetic Beads
Ligand Type Streptavidin
Product Line MagnaBind
Quantity 5 mL
Volume (Metric) 5 mL
Diameter (Metric) 1 to 4 μm
Show More Show Less
What are the differences between Pierce Streptavidin Magnetic Beads and MagnaBind Streptavidin Beads?

Pierce Streptavidin Magnetic Beads are homogeneous, non-aggregating, pre-blocked, iron oxide, superparamagnetic microparticles encapsulated with a proprietary polymer surface (no exposed iron, bead size of approximately 1 (0.5-1.5) µm, concentration 10 mg/mL). Pierce Streptavidin Magnetic Beads can be used in manual or automated applications (using KingFisher 96 and KingFisher Flex instruments, for example).
MagnaBind Streptavidin Beads are also superparamagnetic particles, but are heterogeneous aggregates made of silanized iron oxide (bead size approximately 1-4 µm, concentration 5 mg/mL). MagnaBind Streptavidin Beads can only be used in manual applications.

Can I centrifuge the MagnaBind Streptavidin Beads and Thermo Scientific Streptavidin Magnetic Beads?

No, centrifuging causes the formation of irreversible aggregates which greatly reduces binding capacity.

Can I freeze the MagnaBind Streptavidin Beads and Thermo Scientific Streptavidin Magnetic Beads?

No, freezing or drying will cause the beads to aggregate and lose binding activity. Store the beads only at 4 degrees C.

What are the average sizes of your streptavidin magnetic beads?

Thermo Scientific Streptavidin Magnetic Beads average about 1 micron in daimeter ranging from about 0.5-1.5 microns while MagnaBind Streptavidin Beads are heterogenous averaging 1 - 4 microns.

Are the MagnaBind Streptavidin Beads and Thermo Scientific Streptavidin Magnetic Beads sterile?

No, neither the MagnaBind Streptavidin Beads nor the Thermo Scientific Streptavidin Magnetic Beads are sterilized. However, they are both supplied in solutions containing sodium azide to prevent bacterial growth.

What is the difference between MagnaBind Streptavidin Beads and Thermo Scientific Streptavidin Magnetic Beads?

Thermo Scientific Streptavidin Magnetic Beads are homogenous non-aggregating, pre-blocked, iron oxide, superparamagnetic microparticles encapsulated with proprietary polymer surface (no exposed iron). The MagnaBind Streptavidin Beads on the other hand are also superparamagnetic particles but are hetreogenous aggregates made of silanized iron oxide. The Thermo Scientific Streptavidin Magnetic Beads can be used in manual or automated applications (using Thermo Scientific KingFisher 96 and KingFisher Flex Instruments for example) while MagnaBind Streptavidin Beads can only be used in manual applications.


For Research Use Only. Not for use in diagnostic procedures.