missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Magna RIP™ RNA-Binding Protein Immunoprecipitation Kit

Catalog No. 17700MI
Change view
Click to view available options
Quantity:
12 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17700MI 12 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17700MI Supplier MilliporeSigma™ Supplier No. 17700

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • Magnetic Beads Protein A/G
  • RIP Wash Buffer
  • RIP Lysis Buffer
  • 0.5 M EDTA
  • 10% SDS
  • Salt Solution I
  • Salt Solution II
  • Precipitate Enhancer
  • Normal Mouse IgG
  • Rabbit IgG Purified
  • Protease Inhibitor Cocktail 200X
  • RNase Inhibitor
  • Proteinase K (10mg/mL)
  • Nuclease free water

Requires

Magna Grip Rack 8 well

RIP can be used to identify specific RNA molecules (of many types) associated with specific nuclear or cytoplasmic binding proteins

  • Protein A/G magnetic beads, optimized to bind nucleic acid-protein immune complexes
  • RNAse inhibitors and RNAse-free reagents
  • Negative controls

Specifications

Content And Storage Store components at the temperatures indicated on the labels
For Use With (Application) RNA Binding Protein Immunoprecipitation
Quantity 12 Assays
Stability Stable for 6 months from date of shipment when stored as directed