missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Magna Nuclear RIP™ (Cross-Linked) Nuclear RNA-Binding Protein Immunoprecipitation Kit

Catalog No. 1710520
Change view
Click to view available options
Quantity:
24 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1710520 24 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1710520 Supplier MilliporeSigma™ Supplier No. 1710520

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • 10X Glycine
  • 10X PBS
  • Nuclei Isolation Buffer
  • RIP Cross-Linked Lysis Buffer
  • Protein A/G Magnetic Beads
  • Nuclear RIP Dilution Buffer
  • Low Salt Wash Buffer
  • High Salt Wash Buffer
  • LiCl Wash Buffer
  • TE Buffer
  • RIP Elution Buffer
  • 10% SDS
  • 0.5 M EDTA
  • DNase I (RNase Free)
  • DNase I Supplement
  • DNase I Reaction Buffer
  • Protease Inhibitor Cocktail III, Animal Free
  • RNAse Inhibitor
  • Proteinase K

Requires

  • Antibody of interest for RNA-binding protein immunoprecipitation (RIP)
  • Negative Control Antibody
  • Precipitation Carrier (e.g. Pellet Paint™ Co-Precipitant or linear Acrylamide (5mg/mL) if samples are intended for Next Gen Sequence library preparation
  • RNase A (10mg/mL)
  • Reagents for qRT-PCR Analysis and/or Reagents for RNA-Seq Library Construction

Specially designed to allow the discovery and analysis of a variety of chromatin associated RNAs such as long non-coding RNAs, enhancer RNAs and miRNAs

  • Generates cross-linked chromatin to allow analysis of a variety of chromatin-associated RNAs
  • Flexible, scalable input requirements: Recover RNA from millions of cells or as few as 5,000 cells
  • Magnetic protein A/G bead blend and optimized buffer system results in low backgrounds and high signal-to-noise ratios
  • Suitable for analysis by RT-qPCR or RIP-seq
  • Complete set of reagents and detailed protocol to enable first-time success

Specifications

Content And Storage Store components at the temperatures indicated on the labels
For Use With (Application) RNA Binding Protein Immunoprecipitation (RIP), Protein Interaction Assays
Quantity 24 Assays
Stability Kit components are stable for 6 months from date of shipment when stored as directed