missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Magna ChIP™ Protein G Magnetic Beads
Significantly reduce handling time and mechanical stress on target immunocomplexes
$483.00 - $483.00
Specifications
| Content And Storage | 2°C to 8°C |
|---|---|
| For Use With (Application) | Chromatin immunoprecipitations (ChIP assays) |
| Sample Volume | 1 mL |
| Type | Magnetic Beads |
Description
- Recombinant protein A covalently bound to magnetic beads
- These beads provide users a rapid, reproducible and efficient reagent for collecting immunocomplexes for chromatin immunoprecipitations (ChIP) assays
- Protein A magnetic beads significantly reduce the handling time and mechanical stress on target immunocomplexes
- Presentation: Liquid suspension. Supplied as magnetic bead slurry in phosphate buffered saline, pH 7.4, containing 0.01% Tween™-20 and 0.09% sodium azide.
- Application: Recombinant Protein A covalently bound to magnetic beads for use in chromatin immunoprecipitations (ChIP assays). These protein A beads provide users a more rapid, reproducible & efficient reagent for collecting immunocomplexes vs. agarose beads
Specifications
| 2°C to 8°C | |
| Chromatin immunoprecipitations (ChIP assays) | |
| 1 mL | |
| Magnetic Beads |