missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Magna ChIP™ Protein A Magnetic Beads

Catalog No. 16661MI
Change view
Click to view available options
Quantity:
50 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16661MI 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16661MI Supplier MilliporeSigma™ Supplier No. 16661

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Significantly reduce handling time and mechanical stress on target immunocomplexes

  • Recombinant protein A covalently bound to magnetic beads
  • These beads provide users a rapid, reproducible and efficient reagent for collecting immunocomplexes for chromatin immunoprecipitations (ChIP) assays
  • Protein A magnetic beads significantly reduce the handling time and mechanical stress on target immunocomplexes
  • Presentation: Liquid suspension. Supplied as magnetic bead slurry in phosphate buffered saline, pH 7.4, containing 0.01% Tween™-20 and 0.09% sodium azide.
  • Application: Recombinant Protein A covalently bound to magnetic beads for use in chromatin immunoprecipitations (ChIP assays). These protein A beads provide users a more rapid, reproducible & efficient reagent for collecting immunocomplexes vs. agarose beads

  • Store kit components at the temperatures indicated on the labels and insert until their expiration date
  • Store HeLa Whole Cell Extract at -80°C. Avoid repeated freeze-thaw cycles. Stable at -80°C for 6 months

Specifications

Content And Storage 2°C to 8°C
For Use With (Application) Chromatin immunoprecipitations (ChIP assays)
Quantity 50 Reactions
Sample Volume 1 mL
Type Magnetic Beads