missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Magna ChIP™ G Tissue Kit

Catalog No. 1720000
Change view
Click to view available options
Quantity:
22 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1720000 22 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1720000 Supplier MilliporeSigma™ Supplier No. 1720000

Includes

  • Magnetic Protein G beads
  • ChIP Dilution Buffer
  • Low Salt Wash Buffer
  • High Salt Wash Buffer
  • LiCl Wash Buffer
  • TE Buffer
  • Tissue Lysis Buffer
  • 10X Tissue Stabililizing Solution
  • ChIP Elution buffer (w/o Proteinase K)
  • 10X Glycine
  • 10X PBS
  • 1mm Micro-dissection punch
  • Spin Filters
  • Collection Tubes
  • Bind Reagent A
  • Wash Reagent B
  • Elution Reagent C
  • Protease Inhibitor Cocktail II
  • Proteinase K

Microdissected functionally related populations of cells within a heterogeneous tissue can be analyzed with ease, precision and certainty

  • The kit provides the tools necessary to obtain repeatable, reliable, and site specific tissue biopsies
  • Two modules containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions from tissue samples

Specifications

For Use With (Application) Sample Prep
Quantity 22 Assays