missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ TFS Solution

Catalog No. A45351
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A45351 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A45351 Supplier Applied Biosystems™ Supplier No. A45351

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MagMAX™ TFS Solution is a tissue fragment stability and storage solution developed to ensure the stability of RNA/DNA/proteins during room temperature transportation and/or storage

The MagMAX™ TFS Solution was developed for use with ear tissue biopsies. Applications include genomic DNA sequencing (including CE, microarray, and next-generation sequencing), RNA detection with RT-PCR, and ELISA.

Specifications

Color Clear
pH 7.4 ±0.05
Quantity 1 L

For Veterinary Use Only