Learn More
Abcam Inc Magenta-Gal, Chromogenic beta-galactosidase substrate, 100MG
Catalog No. 502704671
Encompass
MW 408.63 g/mol. Chromogenic substrate for -galactosidase producing a red or magenta insoluble product. Used to determine the presence or absence of a cloned DNA insert in bacteria growing on agar plates. Designed to replace X-Gal in blue-white selection of recombinant bacterial colonies with the lac+ phenotype. Lac+ colonies grown in the presence of Magenta-Gal turn an intense red color, allowing for easy differentiation between lac+ and lac- colonies.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502704671 Supplier Abcam Inc Supplier No. AB146381100MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 408.63 g/mol. Chromogenic substrate for -galactosidase producing a red or magenta insoluble product. Used to determine the presence or absence of a cloned DNA insert in bacteria growing on agar plates. Designed to replace X-Gal in blue-white selection of recombinant bacterial colonies with the lac+ phenotype. Lac+ colonies grown in the presence of Magenta-Gal turn an intense red color, allowing for easy differentiation between lac+ and lac- colonies.
The product is subject to the following: Abcam Restricted Use Statement