Learn More
Mopec Maestro Recirculating to In-House Facility Vented Conversion Kit
Designed to convert recirculating grossing stations into in-house ventilation
Supplier:  Mopec AX110048
 View more versions of this product
Catalog No. 23987020
Only null left
Description
Description
  • Designed to convert recirculating grossing stations into in-house ventilation
  • Bolt-on panel with 8 in duct stubs which allows the grossing station to be connected to the facility ventilation system
  • Includes panel, duct tubing, and clamps
  • 304 stainless steel sheetmetal and hardware
  • Fits the Mopec Maestro and Maestro Encore grossing stations of the accompanying sized unit
Specifications
Chemical Identifiers
Grossing Station
Stainless Steel (304)
Specifications
Grossing Station
Stainless Steel (304)
48 in.
Product Suggestions
Product Suggestions