Learn More
Mopec Maestro Recirculating to In-House Facility Vented Conversion Kit
Catalog No. 23987023
Change view
Click to view available options
Height (English):
48 in.
60 in.
72 in.
96 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English)
23987023 96 in.
23987020 48 in.
23987021 60 in.
23987022 72 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23987023 Supplier Mopec Supplier No. AX110096
Designed to convert recirculating grossing stations into in-house ventilation
  • Designed to convert recirculating grossing stations into in-house ventilation
  • Bolt-on panel with 8 in duct stubs which allows the grossing station to be connected to the facility ventilation system
  • Includes panel, duct tubing, and clamps
  • 304 stainless steel sheetmetal and hardware
  • Fits the Mopec Maestro and Maestro Encore grossing stations of the accompanying sized unit
Specifications
Product Type Grossing Station
Material Stainless Steel (304)
Height (English) 96 in.