Learn More
Mopec Maestro Exhaust Dampening Kit
Catalog No. 23987024
Change view
Click to view available options
Length (English):
48 in.
60 in.
72 in.
96 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
23987024 48 in.
23987025 60 in.
23987026 72 in.
23987027 96 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23987024 Supplier Mopec Supplier No. AX111048
Designed to integrate adjustable exhaust dampeners into your grossing station to help your HVAC/facilities
  • Designed to integrate adjustable exhaust dampeners into your grossing station to help your HVAC/facilities personnel balance the airflow to the grossing stations
  • 304 stainless steel sheetmetal and hardware
  • Fits the Mopec Maestro and Maestro Encore grossing stations of the accompanying sized unit
Specifications
Product Type Grossing Station
Length (English) 48 in.