Learn More
Bioworld Macerozyme R-10, 1 g
Catalog No. 50198983
Encompass
Macerozyme R-10Macerating enzyme from Rhizopus sp. suitable for the isolation of plant cells and can be used in combination with cellulase.Specification:Used in plant protoplast preparation to digest cell wall prior to oranelle isolation.A multi-component enzyme mixture containing approximately: * Cellulase: 0.1 U/mg * Hemicellulase: 0.25 U/mg * Pectinase: 0.5 U/mgSource: Rhizopus sp.Solubility: Soluble in water
Catalog No. 50198983 Supplier Bioworld Supplier No. 215600171
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Macerozyme R-10Macerating enzyme from Rhizopus sp. suitable for the isolation of plant cells and can be used in combination with cellulase.Specification:Used in plant protoplast preparation to digest cell wall prior to oranelle isolation.A multi-component enzyme mixture containing approximately: * Cellulase: 0.1 U/mg * Hemicellulase: 0.25 U/mg * Pectinase: 0.5 U/mgSource: Rhizopus sp.Solubility: Soluble in water