Learn More
Veolia Wts Analytical M9 Conductivity Calibration Standard
Supplier: GE Analytical Instruments STD7702601
Set includes: (1) 1.4 mS/cm (as KCI) in 30 mL specialty Dual Use Conductivity and TOC vial.