Learn More
Simport™ Scientific M480 Disposable Embedding Cassettes in Quickload™ Stacks
Description
- In Quickload™ stacks
- 45° Anterior writing area
- Accept standard metal lids (cat# M481)
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Specifications
| Quantity | 2000/Pk. |
| Color | Orange |
| Material | Acetal |