Learn More
Simport™ Scientific M480 Disposable Embedding Cassettes in Quickload™ Stacks
Catalog No. 22038034
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22038034 White
22038035 Pink
22038036 Green
22038037 Yellow
22038038 Blue
22038039 Peach
22038040 Tan
22038041 Gray
22038042 Lilac
22038043 Orange
22038044 Aqua
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 22038034 Supplier Simport™ Scientific Supplier No. M4802T
Only null left
M480 Disposable Embedding Cassettes in Quickload™ Stacks for holding and identifying tissue samples in processing/embedding procedures.
  • In Quickload™ stacks
  • 45° Anterior writing area
  • Accept standard metal lids (cat# M481)
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Quantity 2000/Pk.
Color White
Material Acetal