missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Xylylenediphosphonic Acid 97.0+%, TCI America™
$105.89 - $319.57
Chemical Identifiers
| CAS | 144052-40-4 |
|---|---|
| Molecular Formula | C8H12O6P2 |
| Molecular Weight (g/mol) | 266.126 |
| MDL Number | MFCD22200402 |
| InChI Key | PYWZCUXISXXGMI-UHFFFAOYSA-N |
| PubChem CID | 19352865 |
| IUPAC Name | [3-(phosphonomethyl)phenyl]methylphosphonic acid |
| SMILES | C1=CC(=CC(=C1)CP(=O)(O)O)CP(=O)(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
X0076200MG
|
TCI America
X0076200MG |
200 mg |
Each for $105.89
|
|
|||||
|
X00761G
|
TCI America
X00761G |
1 g |
Each for $319.57
|
|
|||||
Chemical Identifiers
| 144052-40-4 | |
| 266.126 | |
| PYWZCUXISXXGMI-UHFFFAOYSA-N | |
| [3-(phosphonomethyl)phenyl]methylphosphonic acid |
| C8H12O6P2 | |
| MFCD22200402 | |
| 19352865 | |
| C1=CC(=CC(=C1)CP(=O)(O)O)CP(=O)(O)O |
Specifications
| 144052-40-4 | |
| C8H12O6P2 | |
| 200 mg | |
| PYWZCUXISXXGMI-UHFFFAOYSA-N | |
| [3-(phosphonomethyl)phenyl]methylphosphonic acid | |
| 19352865 | |
| ≥97.0% (HPLC,T) | |
| m-Xylylenediphosphonic Acid |
| 216°C | |
| MFCD22200402 | |
| 3261 | |
| C1=CC(=CC(=C1)CP(=O)(O)O)CP(=O)(O)O | |
| 266.126 | |
| 266.13 | |
| Crystalline Powder |
Safety and Handling
TSCA : No