missing translation for 'onlineSavingsMsg'
Learn More

m-Xylylenediphosphonic Acid 97.0+%, TCI America™

Catalog No. X0076200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X0076200MG 200 mg
X00761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X0076200MG Supplier TCI America Supplier No. X0076200MG
Only null left

Chemical Identifiers

CAS 144052-40-4
Molecular Formula C8H12O6P2
Molecular Weight (g/mol) 266.126
MDL Number MFCD22200402
InChI Key PYWZCUXISXXGMI-UHFFFAOYSA-N
PubChem CID 19352865
IUPAC Name [3-(phosphonomethyl)phenyl]methylphosphonic acid
SMILES C1=CC(=CC(=C1)CP(=O)(O)O)CP(=O)(O)O

Specifications

Melting Point 216°C
Quantity 200 mg
UN Number 3261
Formula Weight 266.13
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material m-Xylylenediphosphonic Acid