missing translation for 'onlineSavingsMsg'
Learn More

m-Tolyltetrazolium Red 98.0+%, TCI America™

Catalog No. T0324100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0324100MG 100 mg
T03241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0324100MG Supplier TCI America Supplier No. T0324100MG

Chemical Identifiers

CAS 88159-25-5
Molecular Formula C20H17ClN4
Molecular Weight (g/mol) 348.834
MDL Number MFCD00059997
InChI Key ARLMHAOZVFIBCT-UHFFFAOYSA-M
Synonym 2,5-Diphenyl-3-(m-tolyl)tetrazolium Chloride
PubChem CID 9798427
IUPAC Name 2-(3-methylphenyl)-3,5-diphenyltetrazol-3-ium;chloride
SMILES CC1=CC(=CC=C1)N2N=C(N=[N+]2C3=CC=CC=C3)C4=CC=CC=C4.[Cl-]

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 348.83
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material m-Tolyltetrazolium Red