Learn More
m-Tolylacetic acid, 99%
CAS: 621-36-3 | C9H10O2 | 150.18 g/mol
Supplier: Thermo Scientific Chemicals A1774330
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water.
Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
Specifications
| m-Tolylacetic acid | |
| 62°C to 66°C | |
| MFCD00004340 | |
| 2043549 | |
| Soluble in water. | |
| CC1=CC=CC(CC(O)=O)=C1 | |
| 150.18 | |
| 150.18 |
| 621-36-3 | |
| C9H10O2 | |
| 250 g | |
| 3-methylphenylacetic acid, m-tolylacetic acid, 2-m-tolyl acetic acid, 2-3-methylphenyl acetic acid, 3-tolylacetic acid, m-methylphenylacetic acid, 3-methylphenyl acetic acid, m-tolylaceticacid, benzeneacetic acid, 3-methyl, methyll 3-methylphenylacetate | |
| GJMPSRSMBJLKKB-UHFFFAOYSA-N | |
| 2-(3-methylphenyl)acetic acid | |
| 12121 | |
| 99% |
Chemical Identifiers
| 621-36-3 | |
| 150.18 | |
| GJMPSRSMBJLKKB-UHFFFAOYSA-N | |
| 12121 | |
| CC1=CC=CC(CC(O)=O)=C1 |
| C9H10O2 | |
| MFCD00004340 | |
| 3-methylphenylacetic acid, m-tolylacetic acid, 2-m-tolyl acetic acid, 2-3-methylphenyl acetic acid, 3-tolylacetic acid, m-methylphenylacetic acid, 3-methylphenyl acetic acid, m-tolylaceticacid, benzeneacetic acid, 3-methyl, methyll 3-methylphenylacetate | |
| 2-(3-methylphenyl)acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-683-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only