missing translation for 'onlineSavingsMsg'
Learn More

m-Tolyl Isocyanate 99.0+%, TCI America™

Catalog No. p-7136407
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T072125ML 25 mL
T0721500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T072125ML Supplier TCI America Supplier No. T072125ML
Only null left

Chemical Identifiers

CAS 621-29-4
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00002021
InChI Key CPPGZWWUPFWALU-UHFFFAOYSA-N
Synonym m-tolyl isocyanate, benzene, 1-isocyanato-3-methyl, meta-tolyl isocyanate, m-tolylisocyanate, m-isocyanatotoluene, 3-methylphenyl isocyanate, 3-tolyl isocyanate, 3-methylphenylisocyanate, 1-isocyanato-3-methyl-benzene, m-methylphenyl isocyanate
PubChem CID 69303
IUPAC Name 1-isocyanato-3-methylbenzene
SMILES CC1=CC=CC(=C1)N=C=O

Specifications

Boiling Point 96°C
Quantity 25 mL
UN Number 2206
Formula Weight 133.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material m-Tolyl Isocyanate
EINECSNumber (3)-3238
TSCA Yes
Recommended Storage Refrigerator