missing translation for 'onlineSavingsMsg'
Learn More

m-Toluoylacetonitrile, 97%

Catalog No. p-7081811
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
AAA1196503 1 g
AAA1196506 5 g
2 options

Catalog No. AAA1196503

Supplier: Thermo Scientific Chemicals A1196503

Only null left

CAS: 53882-81-8 | C10H9NO | 159.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53882-81-8
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00067922
InChI Key IVLKDYOTZMFMLO-UHFFFAOYSA-N
Synonym 3-methylbenzoylacetonitrile, 3-3-methylphenyl-3-oxopropanenitrile, 3-toluoylacetonitrile, m-toluoylacetonitrile, 3-oxo-3-m-tolyl propanenitrile, 3-oxo-3-m-tolylpropanenitrile, pubchem12073, maybridge1_004502, acmc-1alb6, 3-m-tolyl-3-oxo-propanenitrile
PubChem CID 143105
IUPAC Name 3-(3-methylphenyl)-3-oxopropanenitrile
SMILES CC1=CC=CC(=C1)C(=O)CC#N

Specifications

Melting Point 71°C to 75°C
Quantity 1 g
Beilstein 2717335
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material m-Toluoylacetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only