missing translation for 'onlineSavingsMsg'
Learn More

m-Tolunitrile 98.0+%, TCI America™

Catalog No. T030525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T030525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T030525ML Supplier TCI America Supplier No. T030525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 620-22-4
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.151
MDL Number MFCD00001808
InChI Key BOHCMQZJWOGWTA-UHFFFAOYSA-N
Synonym m-tolunitrile, 3-cyanotoluene, benzonitrile, 3-methyl, m-cyanotoluene, 3-tolunitrile, m-toluonitrile, m-tolylnitrile, m-toluenenitrile, m-methylbenzonitrile, m-tolynitrile
PubChem CID 12104
IUPAC Name 3-methylbenzonitrile
SMILES CC1=CC=CC(=C1)C#N

Specifications

Color Colorless
Boiling Point 210°C
Quantity 25 mL
Formula Weight 117.15
Physical Form Liquid
Chemical Name or Material m-Tolunitrile
EINECSNumber (3)-1804
RTECSNumber XV0525000
TSCA Yes