missing translation for 'onlineSavingsMsg'
Learn More

m-Tolunitrile, 99%

Catalog No. AC158010050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158010050 5 g Glass bottle
AC158011000 100 g Glass bottle
AC158015000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC158010050 Supplier Thermo Scientific Chemicals Supplier No. 158010050
Only null left

CAS: 620-22-4 | C8H7N | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 620-22-4
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.15
MDL Number MFCD00001808
InChI Key BOHCMQZJWOGWTA-UHFFFAOYSA-N
Synonym m-tolunitrile, 3-cyanotoluene, benzonitrile, 3-methyl, m-cyanotoluene, 3-tolunitrile, m-toluonitrile, m-tolylnitrile, m-toluenenitrile, m-methylbenzonitrile, m-tolynitrile
PubChem CID 12104
IUPAC Name 3-methylbenzonitrile
SMILES CC1=CC=CC(=C1)C#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -23°C
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 99°C to 101°C (20.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5246 to 1.5266
Linear Formula CH3C6H4CN
Quantity 5 g
Beilstein 09, 477
Specific Gravity 0.97
Solubility Information Solubility in water: .. Other solubilities: solunle in methanol,ethanol,dioxane
Formula Weight 117.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material m-Tolunitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective cloth
EINECSNumber 210-631-5

RUO – Research Use Only