Learn More
m-Toluenesulfonyl chloride, 98%
CAS: 1899-93-0 | C7H7ClO2S | 190.641 g/mol
Supplier: Thermo Scientific Chemicals A1545814
Description
Reactant involved in they synthesis of biologically active molecules including small molecule ligands for the Stat3 SH2 domain, aryldisulfonamides with antibacterial activity, quinazoline analogs for the treatment of Gaucher disease, 17β-HSD2 inhibitors for the treatment of osteoporosis, isoindoline-5-propenehydroxamates for use as histone deactylase inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReactant involved in they synthesis of biologically active molecules including small molecule ligands for the Stat3 SH2 domain, aryldisulfonamides with antibacterial activity, quinazoline analogs for the treatment of Gaucher disease, 17β-HSD2 inhibitors for the treatment of osteoporosis, isoindoline-5-propenehydroxamates for use as histone deactylase inhibitors.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents.
Specifications
| m-Toluenesulfonyl chloride | |
| 1.314 | |
| 107°C (224°F) | |
| C7H7ClO2S | |
| MFCD00051747 | |
| UN3265 | |
| Moisture sensitive | |
| Reacts with water. | |
| CC1=CC(=CC=C1)S(=O)(=O)Cl | |
| 190.641 | |
| 190.65 |
| 1899-93-0 | |
| 252°C to 253°C | |
| Acrid | |
| 1.548 | |
| 25 g | |
| 2087865 | |
| m-toluenesulfonyl chloride, 3-methylbenzene-1-sulfonyl chloride, 3-toluenesulfonyl chloride, 3-methyl-benzenesulfonyl chloride, 3-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 3-methyl, 3-toluenesulphonyl chloride, 3-methylphenylsulfonyl chloride, pubchem5624, m-tolylsulfonyl chloride | |
| KFPMLWUKHQMEBU-UHFFFAOYSA-N | |
| 3-methylbenzenesulfonyl chloride | |
| 2734595 | |
| 98% |
Chemical Identifiers
| 1899-93-0 | |
| 190.641 | |
| KFPMLWUKHQMEBU-UHFFFAOYSA-N | |
| 2734595 | |
| CC1=CC(=CC=C1)S(=O)(=O)Cl |
| C7H7ClO2S | |
| MFCD00051747 | |
| m-toluenesulfonyl chloride, 3-methylbenzene-1-sulfonyl chloride, 3-toluenesulfonyl chloride, 3-methyl-benzenesulfonyl chloride, 3-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 3-methyl, 3-toluenesulphonyl chloride, 3-methylphenylsulfonyl chloride, pubchem5624, m-tolylsulfonyl chloride | |
| 3-methylbenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only