Learn More
m-Toluenesulfonic acid monohydrate, 97%
CAS: 312619-56-0 | C7H10O4S | 190.213 g/mol
Supplier: Thermo Scientific Chemicals L0341914
Description
m-Toluenesulfonic acid monohydrate is used in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsm-Toluenesulfonic acid monohydrate is used in chemical research.
Solubility
Fully miscible in water.
Notes
Hygroscopic. Store away from strong bases, moisture. Incompatible materials are oxidizing agents, bases.
Specifications
| m-Toluenesulfonic acid monohydrate | |
| 165°C (0.1 mmHg) | |
| 1.527 | |
| 25 g | |
| 2208421 | |
| Fully miscible in water. | |
| CC1=CC(=CC=C1)S(=O)(=O)O.O | |
| 190.213 | |
| 190.22 (172.20 Anhydrous) |
| 312619-56-0 | |
| C7H10O4S | |
| MFCD00167130 | |
| UN2586 | |
| 3-methylbenzenesulfonic acid hydrate, m-toluenesulfonic acid monohydrate, m-toluenesulfonic acid hydrate, acmc-20akdl, m-toluenesulfonic acid 1-hydrate, 3-toluenesulfonic acid 1-hydrate, 3-methylbenzenesulfonic acid hydrate 1:x, benzenesulfonicacid, 3-methyl-, hydrate 1:1 | |
| GHLYGGIKYFLYTH-UHFFFAOYSA-N | |
| 3-methylbenzenesulfonic acid;hydrate | |
| 53249652 | |
| 97% |
Chemical Identifiers
| 312619-56-0 | |
| 190.213 | |
| GHLYGGIKYFLYTH-UHFFFAOYSA-N | |
| 53249652 | |
| CC1=CC(=CC=C1)S(=O)(=O)O.O |
| C7H10O4S | |
| MFCD00167130 | |
| 3-methylbenzenesulfonic acid hydrate, m-toluenesulfonic acid monohydrate, m-toluenesulfonic acid hydrate, acmc-20akdl, m-toluenesulfonic acid 1-hydrate, 3-toluenesulfonic acid 1-hydrate, 3-methylbenzenesulfonic acid hydrate 1:x, benzenesulfonicacid, 3-methyl-, hydrate 1:1 | |
| 3-methylbenzenesulfonic acid;hydrate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYL SULFONIC ACIDS, LIQUID
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only