missing translation for 'onlineSavingsMsg'
Learn More

m-Tolualdehyde 2,4-Dinitrophenylhydrazone 98.0+%, TCI America™

Catalog No. p-7136782
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14481G 1 g
T144810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14481G Supplier TCI America Supplier No. T14481G
Only null left

Chemical Identifiers

CAS 2880-05-9
Molecular Formula C14H12N4O4
Molecular Weight (g/mol) 300.27
MDL Number MFCD00191585
InChI Key PQBRXQUGVASQBU-UHFFFAOYSA-N
Synonym m-tolualdehyde 2,4-dinitrophenylhydrazone, m-tolualdehyde, 2,4-dinitrophenyl hydrazone, benzaldehyde, 3-methyl-, 2,4-dinitrophenyl hydrazone, 3'-methylbenzaldehyde 2,4-dinitrophenylhydrazone, m-tolualdehyd-2,4-dinitrophenylhydrazone, m-tolualdehyde 2,4-dinitro-phenylhydrazone, 3-methylbenzaldehyde 2,4-dinitrophenyl hydrazone #, 1-2,4-dinitrophenyl-2-3-methylbenzylidene hydrazine, n-e-3-methylphenyl methylideneamino-2,4-dinitroaniline, m-tolualdehyde 2,4-dinitrophenylhydrazone, environmental standard
PubChem CID 9604108
IUPAC Name N-[(E)-(3-methylphenyl)methylideneamino]-2,4-dinitroaniline
SMILES CC1=CC=CC(=C1)C=NNC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 213°C
Quantity 1 g
UN Number 1325
Formula Weight 300.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material m-Tolualdehyde 2,4-Dinitrophenylhydrazone