missing translation for 'onlineSavingsMsg'
Learn More

m-Phosphoric Acid (Cryst. or Flakes/Certified ACS), Fisher Chemical™

Catalog No. p-32751
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A280100 100 g Glass Bottle
A280500 500 g Glass Bottle
A2803 3 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A280100 Supplier Thermo Fisher Scientific Supplier No. FLA280100
Only null left

Mixture of meta-Phosphoric Acid and Sodium Metaphosphate

Chemical Identifiers

CAS 37267-86-0 , 50813-16-6
Molecular Formula HO3P
Molecular Weight (g/mol) 79.979
MDL Number MFCD00011351
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
Synonym metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
PubChem CID 3084658
SMILES OP(=O)=O

Specifications

Density 2.4g/cm³
Heavy Metals (as Pb) 0.005% max.
Identification Pass Test
Packaging Glass Bottle
Assay Percent Range 33.5 to 36.5 %
Quantity 100 g
Physical Form Solid
Percent Purity 33.5 to 36.5%
Grade Certified ACS
Chemical Name or Material m-Phosphoric Acid
DOTInformation DOT Class 8, : Corrosive