missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich m-Phenylenediamine
Description
m-Phenylenediamine
Specifications
Specifications
| CAS | 108-45-2 |
| Melting Point | 64°C to 66°C |
| Boiling Point | 282°C to 284°C |
| Molecular Formula | C6H8N2 |
| Linear Formula | C6H4-1,3-(NH2)2 |
| MDL Number | MFCD00007799 |
| Quantity | 15 kg |
| Synonym | 1,3-Benzenediamine; 1,3-Diaminobenzene; 1,3-Phenylenediamine; MPDA |
| Molecular Weight (g/mol) | 108.14 |
| Percent Purity | 99% |