missing translation for 'onlineSavingsMsg'
Learn More

m-Hydroxycinnamic acid, 99%, predominantly trans, Thermo Scientific™

Chemical Identifiers

CAS 14755-02-3
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00004390
InChI Key KKSDGJDHHZEWEP-SNAWJCMRSA-N
Synonym 3-hydroxycinnamic acid, 3-coumaric acid, m-coumaric acid, m-hydroxycinnamic acid, 3-3-hydroxyphenyl acrylic acid, e-3-3-hydroxyphenyl acrylic acid, trans-3-hydroxycinnamic acid, trans-3-hydroxycinnamate, cinnamic acid, m-hydroxy, 2e-3-3-hydroxyphenyl prop-2-enoic acid
PubChem CID 637541
ChEBI CHEBI:32357
IUPAC Name (E)-3-(3-hydroxyphenyl)prop-2-enoic acid
SMILES C1=CC(=CC(=C1)O)C=CC(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC121080250
SDS View Documents
Thermo Scientific Chemicals
121080250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

14755-02-3
164.16
KKSDGJDHHZEWEP-SNAWJCMRSA-N
637541
(E)-3-(3-hydroxyphenyl)prop-2-enoic acid
C9H8O3
MFCD00004390
3-hydroxycinnamic acid, 3-coumaric acid, m-coumaric acid, m-hydroxycinnamic acid, 3-3-hydroxyphenyl acrylic acid, e-3-3-hydroxyphenyl acrylic acid, trans-3-hydroxycinnamic acid, trans-3-hydroxycinnamate, cinnamic acid, m-hydroxy, 2e-3-3-hydroxyphenyl prop-2-enoic acid
CHEBI:32357
C1=CC(=CC(=C1)O)C=CC(=O)O

Specifications

14755-02-3
100.0
99%
C9H8O3
MFCD00004390
10, 294
KKSDGJDHHZEWEP-SNAWJCMRSA-N
(E)-3-(3-hydroxyphenyl)prop-2-enoic acid
637541
164.16
m-Hydroxycinnamic acid, Predominantly trans
98.5
Authentic
Glass Bottle
HOC6H4CH=CHCO2H
25g
3-hydroxycinnamic acid, 3-coumaric acid, m-coumaric acid, m-hydroxycinnamic acid, 3-3-hydroxyphenyl acrylic acid, e-3-3-hydroxyphenyl acrylic acid, trans-3-hydroxycinnamic acid, trans-3-hydroxycinnamate, cinnamic acid, m-hydroxy, 2e-3-3-hydroxyphenyl prop-2-enoic acid
C1=CC(=CC(=C1)O)C=CC(=O)O
164.16
CHEBI:32357
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only