Learn More
Lifeline Cell Technology M FIBS HUXF LIFELINE KT
Catalog No. 50311917
Encompass
FibroLife Xeno-Free (500 mL) is optimized for culture of fibroblasts in 2% human serum environment at growth rates equal to or greater than comparable media supplemented with 2-10% FBS. Complete frozen medium, up to 2 year shelf life.
Catalog No. 50311917 Supplier Lifeline Cell Technology Supplier No. LM0013
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
FibroLife Xeno-Free (500 mL) is optimized for culture of fibroblasts in 2% human serum environment at growth rates equal to or greater than comparable media supplemented with 2-10% FBS. Complete frozen medium, up to 2 year shelf life.