missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Cresol Purple TS, 0.1% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 240516
Specifications
| m-Cresol Purple | |
| 62625-31-4,7732-18-5 | |
| 100.0,0.1 | |
| 0.1% W/V | |
| Natural Poly Bottle | |
| Red/Orange | |
| 500 mL |
| Test Solution | |
| 98.0,0.1 | |
| 0.1% (w/v) | |
| Indicator | |
| Aqueous Solution | |
| 6.85 to 7.05 | |
| Liquid |
Chemical Identifiers
| 62625-31-4 |