missing translation for 'onlineSavingsMsg'
Learn More

m-Cresol Purple

CAS: 2303-01-7 | C21H18O5S | 382.43 g/mol

Supplier:  Thermo Scientific Chemicals B2433814

 View more versions of this product

Catalog No. AAB2433814


Only null left
Explore available promotions

Description

Description

m-Cresol Purple acts as a pH indicator dye. It plays an important role in spectrophotometric pH measurements. It operates in the pH ranges 1.2 - 2.8 (red to yellow) and 7.4 - 9.0 (yellow to violet). Further, it is employed to stain epithelial cells and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Cresol Purple acts as a pH indicator dye. It plays an important role in spectrophotometric pH measurements. It operates in the pH ranges 1.2 - 2.8 (red to yellow) and 7.4 - 9.0 (yellow to violet). Further, it is employed to stain epithelial cells and proteins.

Solubility
Soluble in water and sodium hydroxide solution. Slightly soluble in alcohol.

Notes
Transition interval (acid): pH 1.2 (red) to pH 2.8 (yellow); (alkaline) pH 7.4 (yellow) to pH 9.0 (purple)Incompatible with strong oxidizing agents.
Specifications

Specifications

m-Cresol Purple
C21H18O5S
350314
m-Cresolsulfonephthalein
OLQIKGSZDTXODA-UHFFFAOYSA-N
4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
73030
25 g
2303-01-7
MFCD00005871
14,2577
Soluble in water and sodium hydroxide solution. Slightly soluble in alcohol.
CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C
382.43
382.44

Chemical Identifiers

2303-01-7
382.43
OLQIKGSZDTXODA-UHFFFAOYSA-N
73030
CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C
C21H18O5S
MFCD00005871
m-Cresolsulfonephthalein
4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 218-960-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only